Learn More
Thermo Scientific™ Nalgene™ Torque Wrenches
Assure accurate torque readings when applying closures with Nalgene Torque Wrench Fittings.
$666.50 - $701.00
Specifications
For Use With (Equipment) Nalgene Torque Wrench Fittings (Series 2195)
Type Torque Wrench
View More Specs
Products 2
Catalog Number Mfr. No. Drive Size Price Quantity & Availability  
Catalog Number Mfr. No. Drive Size Price Quantity & Availability  
03-313-50
View Documents
Thermo Scientific™
21956169
1/4 in.
Each for $666.50
Add to cart
02-923-135
View Documents
Thermo Scientific™
21956177
3/8 in.
Case for $701.00
Add to cart
Description
Description
Assure accurate torque readings when applying closures with Thermo Scientific™ Nalgene Torque Wrench Fittings. Proper torquing of Nalgene closures is critical to leakproof† sealing.
These wrenches can be used to confirm capping machine settings, on-line inspection and to check torques. Must be used with Nalgene torque wrench fittings.
Ordering Information:
Order fittings that match closure resin.
Compatible with:
Thermo Scientific Nalgene Torque Wrench Fittings (Series 2195) † The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. B) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Warranty: 90 days
Specifications
Specifications
Nalgene Torque Wrench Fittings (Series 2195)
Torque Wrench
SDS
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.