Learn More
Thermo Scientific Nalgene Unwire Half-Racks
Space-efficient half-racks are ideal for use in incubators, refrigerators, freezers and under fume hoods.
$17.50 - $92.00
Catalog Number | Mfr. No. | Color | Holds | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Holds | Price | Quantity & Availability | ||||
S478321
![]() |
Thermo Scientific
59720013/EMD |
White | Tubes (13 mm) |
Each for $62.25
Case of 8 EA for $497.00 |
|
S478322
![]() ![]() |
Thermo Scientific
59720313/EMD |
Blue | Tubes (13 mm) |
Each for $17.50
Case of 8 EA for $137.00 |
|
S478323
![]() |
Thermo Scientific
59720016/EMD |
White | Tubes (16 mm) |
Each for $92.00
Case of 8 EA for $725.00 |
|
S478324
![]() |
Thermo Scientific
59720316/EMD |
Blue | Tubes (16 mm) |
Each for $92.00
Case of 8 EA for $725.00 |
|
Description
Fit easily in ice or water baths without floating.
- Made of durable ResMere performance plastic
- Withstand most lab chemicals and temperatures up to 121°C (249.8°F)
- Repeatedly autoclavable
- Each rack holds 36 tubes
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Plastic | |
36 | |
Test Tube Half Rack |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.