Thermo Scientific™ Nalgene™ Vented Unitary Right-to-Know LDPE Wash Bottles Available on GSA/VA Contract for Federal Government customers only.

Keep hazardous volatile liquids off the lab bench. Unitary wash bottles feature a unique vent system, and eliminate draw tube assembly and associated risk of contamination.

Manufacturer: Thermo Scientific™ 24360251

 View more versions of this product

Catalog No. 03-409-19A

  Check Availability
Add to Cart

Disclaimers:

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement. The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. B) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.


    Specifications

    Item Description Wash Bottle Unitary™ RTK Acetone LDPE Red 250mL
    Capacity (Metric) 250mL
    Capacity (English) 8 oz.,8 oz.
    Certifications/Compliance Meet OSHA requirements for workplace chemical container labeling.
    Chemical Label Acetone
    Description RTK Wash Bottle
    Material LDPE
    No. per Pack 4
    Capacity 8 oz., 250mL
    Unitary Yes
    Closure Color Red
    Closure Material Polypropylene
    No. per Case 36

    Keep hazardous volatile liquids off the lab bench with Thermo Scientific™ Nalgene™ Vented Unitary Right-To-Know LDPE Wash Bottles, featuring a unique vent system. Unitary bottle prevents contamination when filling the bottle; no draw tube assembly to pick up contamination off the lab bench. A strong, steady stream of liquid is dispensed when the wash bottle is squeezed. Marked with four color, NFPA diamonds, GHS compliant hazard symbols, and right-to-know chemical information. Choose from seven chemical markings.

    • Bottle and tubulation molded in one piece to prevent chemical splash when removing closure and contamination when filling
    • Unique 38mm colored vented closure designed to prevent dripping
    • Sturdy polypropylene closures (HDPE on Sodium Hypochloride bottles)
    • Color-coded bar corresponds to closure, preventing cross-contamination
    • Prelabeled with ICS name and formula, as well as NFPA symbols for chemical handling, hazard, care and storage of six commonly-used chemicals
    • Leakproof†

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit