Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nalidixic acid, 50mg/mL in water, sterile-filtered

Click to view available options
Quantity:
1mL
5 x 1mL
389-08-2
C12H12N2O3
232.239
MFCD00006884
MHWLWQUZZRMNGJ-UHFFFAOYSA-N
nalidixic acid,nalidixin,nevigramon,nalidixate,uronidix,innoxalon,nalidixan,nalitucsan,sicmylon,unaserus
4421
CHEBI:100147
1-ethyl-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid
CCN1C=C(C(=O)C2=C1N=C(C=C2)C)C(=O)O
389-08-2
C12H12N2O3
232.239
MFCD00006884
MHWLWQUZZRMNGJ-UHFFFAOYSA-N
nalidixic acid,nalidixin,nevigramon,nalidixate,uronidix,innoxalon,nalidixan,nalitucsan,sicmylon,unaserus
4421
CHEBI:100147
1-ethyl-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid
CCN1C=C(C(=O)C2=C1N=C(C=C2)C)C(=O)O
Applications
Nalidixic acid is an inhibitor of bacterial DNA polymerase (DNA gyrase) and avian myeloblastoma virus reverse transcriptase. It is used to modulate (inhibit) bacterial DNA gyrase-dependent processes such as DNA polymerization, (ATP-dependent)t DNA supercoiling, and chromosome fragmentation.

Solubility
Soluble in chloroform (50 mg/ml), ethanol (0.9 mg/ml) at 23°C, methanol (1.3 mg/ml) at 23°C, toluene (1.6 mg/ml) at 23°C, DMSO (≥ 6 mg/ml), and solution of carbonates. Fully miscible in water.

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
Quantity 1mL
Physical Form Liquid
Chemical Name or Material Nalidixic acid, 50mg/mL in water, sterile-filtered

RTECSNumber : QN2885000

storageNote1 : -30° to -10°C