Learn More
Nalidixic acid, 99.5%, Thermo Scientific Chemicals
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• Nalixidic acid is a synthetic quinolone that is a monocarboxylic acid composed of 1,8-naphthyridine substituted with carboxylic acid, ethyl, and methyl groups
• Nalixidic acid is a DNA gyrase inhibitor that prevents DNA synthesis and subsequent cell divisions
Applications
• Nalixidic acid is primarily effective against gram-negative bacteria
• It may be useful for bacterial resistance studies and laboratory research on drug allergies and cross-reactivity
• Nalixidic acid is suitable for high-performance liquid chromatography (HPLC) and related biochemistry laboratory applications
Specifications
CAS Min % | 99.4 |
CAS Max % | 100.0 |
Melting Point | 225.0°C to 231.0°C |
Color | White to Yellow |
Infrared Spectrum | Authentic |
Assay Percent Range | 99.5% |
Packaging | Glass bottle |
Quantity | 5 g |
Beilstein | 25, V,7, 384 |
Merck Index | 15, 6444 |
Show More |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement May cause an allergic skin reaction. May cause allergy or asthma symptoms or breathing difficulties if inhaled. |
Precautionary Statement | GHS P Statement Avoid breathing dust/fume/gas/mist/vapors/spray. If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician. Wear eye protection/face protection. |
EINECSNumber | 206-864-7 |
RTECSNumber | QN2885000 |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.