Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nalidixic acid sodium salt, 50mg/mL in chloroform, sterile-filtered

Click to view available options
Quantity:
1mL
5 x 1mL
3374-05-8
C12H11N2NaO3
254.22
MFCD00064376
ROKRAUFZFDQWLE-UHFFFAOYSA-M
nalidixic acid sodium salt,baktogram,nalidixic acid sodium,sodium nalidixate,chemiurin,dixilina,nalidixate sodium anhydrous,nalidixate sodium,nalidixate sodium anhydrous usan,1-ethyl-1,4-dihydro-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid sodium salt
3864541
sodium;1-ethyl-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylate
CCN1C=C(C(=O)C2=C1N=C(C=C2)C)C(=O)[O-].[Na+]
3374-05-8
C12H11N2NaO3
254.22
MFCD00064376
ROKRAUFZFDQWLE-UHFFFAOYSA-M
nalidixic acid sodium salt,baktogram,nalidixic acid sodium,sodium nalidixate,chemiurin,dixilina,nalidixate sodium anhydrous,nalidixate sodium,nalidixate sodium anhydrous usan,1-ethyl-1,4-dihydro-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylic acid sodium salt
3864541
sodium;1-ethyl-7-methyl-4-oxo-1,8-naphthyridine-3-carboxylate
CCN1C=C(C(=O)C2=C1N=C(C=C2)C)C(=O)[O-].[Na+]
Quantity 1mL
UN Number UN1888
Physical Form Liquid
Chemical Name or Material Nalidixic acid sodium salt, 50mg/mL in chloroform, sterile-filtered

GHS H Statement
H334-H351-H373-H302-H315-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Causes skin irritation.
May cause an allergic skin reaction.

GHS P Statement
P260-P285-P261-P342+P311-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
In case of inadequate ventilation wear respiratory protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: CHLOROFORM

RTECSNumber : QN2885000

storageNote1 : -30° to -10°C