Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NLRP6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521022
Description
A suggested positive control is human heart tissue lysate. PA5-21022 can be used with blocking peptide PEP-1136.
NALP proteins include the apoptosis regulator APAF1 (apoptotic protease activating factor 1) and mammalian NOD-LRR proteins and are thought to be involved in inflammation and reproduction. NALP6, also known as PYPAF5, is highly expressed in granulocytes and T cells and co-localizes with the PYRIN-CARD protein ASC. Co-expression of NALP6 with ASC results in a synergistic activation of NF-kappa-B and activated caspase-1-dependent cytokine processing, suggesting that NALP6 functions in immune cells to coordinate the transduction of pro-inflammatory signals.
Specifications
NLRP6 | |
Polyclonal | |
Unconjugated | |
NLRP6 | |
AI504961; angiotensin II/vasopressin receptor; angiotensin/vasopressin receptor; AVR; CLR11.4; dual angiotensin II/vasopressin receptor; NACHT, leucine rich repeat and PYD containing 6; NACHT, LRR and PYD domains-containing protein 6; NACHT/LRR/pyrin domain-containing protein 6; NALP6; Navr; Navr/Avr; NLR family pyrin domain containing 6; NLR family, pyrin domain containing 6; Nlrp6; Non-angiotensin-vasopressin receptor; Non-AVR; nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domain containing 6; PAN3; Pypaf5; PYRIN-containing APAF1-like protein 5; PYRIN-containing APAF1-like protein 5-like | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
171389 | |
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
Liquid |
Immunohistochemistry, Western Blot | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P59044 | |
NLRP6 | |
A 16 amino acid peptide near the amino terminus of human NALP6. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction