Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nalpha-1-(4,4-Dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl-Nepsilon-Fmoc-L-lysine, 98%

Click to view available options
Quantity:
250mg
1g
156648-40-7
C31H36N2O6
532.637
MFCD00467662
CNDRLQTWKWIHEP-VWLOTQADSA-N
dde-lys fmoc-oh,dde-l-lys fmoc-oh,dde-lysine fmoc-oh,s-6-9h-fluoren-9-yl methoxy carbonyl amino-2-1-4,4-dimethyl-2,6-dioxocyclohexylidene ethyl amino hexanoic acid,n-a-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene ethyl-n-e-fmoc-l-lysine,n-alpha-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene ethyl-n-epsilon-fmoc-l-lysine,2s-2-1-4,4-dimethyl-2,6-dioxocyclohexylidene ethyl amino-6-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid,ambotzdaa1015,n-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene ethyl-n'-fmoc-l-lysine,n-,a-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene ethyl-n-,a-fmoc-l-lysine
51340473
(2S)-2-[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)ethylamino]-6-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
CC(=C1C(=O)CC(CC1=O)(C)C)NC(CCCCNC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)O
156648-40-7
C31H36N2O6
532.637
MFCD00467662
CNDRLQTWKWIHEP-VWLOTQADSA-N
dde-lys fmoc-oh,dde-l-lys fmoc-oh,dde-lysine fmoc-oh,s-6-9h-fluoren-9-yl methoxy carbonyl amino-2-1-4,4-dimethyl-2,6-dioxocyclohexylidene ethyl amino hexanoic acid,n-a-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene ethyl-n-e-fmoc-l-lysine,n-alpha-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene ethyl-n-epsilon-fmoc-l-lysine,2s-2-1-4,4-dimethyl-2,6-dioxocyclohexylidene ethyl amino-6-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid,ambotzdaa1015,n-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene ethyl-n'-fmoc-l-lysine,n-,a-1-4,4-dimethyl-2,6-dioxocyclohex-1-ylidene ethyl-n-,a-fmoc-l-lysine
51340473
(2S)-2-[1-(4,4-dimethyl-2,6-dioxocyclohexylidene)ethylamino]-6-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
CC(=C1C(=O)CC(CC1=O)(C)C)NC(CCCCNC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)O
Formula Weight 532.64
Physical Form Powder
Percent Purity 98%
Color White
Quantity 1g
Chemical Name or Material Nalpha-1-(4,4-Dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl-Nepsilon-Fmoc-L-lysine

storageNote1 : -30° to -10°C