Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-tetradecanoyl-L-lysine, TCI America™

Click to view available options
Quantity:
1g
5g
1128181-23-6
C35H50N2O5
578.794
PNXSYZSEFDFTDZ-YTTGMZPUSA-N
Nalpha-Fmoc-Nepsilon-tetradecanoyl-L-lysine, Fmoc-Lys(myristoyl)-OH, Fmoc-Lys(Myr)-OH
25231984
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-(tetradecanoylamino)hexanoic acid
CCCCCCCCCCCCCC(=O)NCCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
1128181-23-6
C35H50N2O5
578.794
PNXSYZSEFDFTDZ-YTTGMZPUSA-N
Nalpha-Fmoc-Nepsilon-tetradecanoyl-L-lysine, Fmoc-Lys(myristoyl)-OH, Fmoc-Lys(Myr)-OH
25231984
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-(tetradecanoylamino)hexanoic acid
CCCCCCCCCCCCCC(=O)NCCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
Melting Point 140°C
Quantity 1g
Color White
Formula Weight 578.79
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (HPLC,N)
Chemical Name or Material Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nepsilon-tetradecanoyl-L-lysine