Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nalpha-Acetyl-Nepsilon-Boc-L-lysine, 95%

Click to view available options
Quantity:
5g
25g
100g
23500-04-1
C13H24N2O5
288.344
MFCD00236761
NZAMQYCOJSWUAO-JTQLQIEISA-N
ac-lys boc-oh,s-2-acetamido-6-tert-butoxycarbonyl amino hexanoic acid,acetyl-nepsilon-boc-l-lysine,2s-6-tert-butoxycarbonyl amino-2-acetamidohexanoic acid,n~2~-acetyl-n~6~-tert-butoxycarbonyl-l-lysine,s-2-acetamido-6-tert-butoxycarbonyl amino hexanoicacid
7019610
(2S)-2-acetamido-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
CC(=O)NC(CCCCNC(=O)OC(C)(C)C)C(=O)O
23500-04-1
C13H24N2O5
288.344
MFCD00236761
NZAMQYCOJSWUAO-JTQLQIEISA-N
ac-lys boc-oh,s-2-acetamido-6-tert-butoxycarbonyl amino hexanoic acid,acetyl-nepsilon-boc-l-lysine,2s-6-tert-butoxycarbonyl amino-2-acetamidohexanoic acid,n~2~-acetyl-n~6~-tert-butoxycarbonyl-l-lysine,s-2-acetamido-6-tert-butoxycarbonyl amino hexanoicacid
7019610
(2S)-2-acetamido-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
CC(=O)NC(CCCCNC(=O)OC(C)(C)C)C(=O)O
Formula Weight 288.34
Physical Form Powder
Percent Purity 95%
Color White
Quantity 25g
Chemical Name or Material Nalpha-Acetyl-Nepsilon-Boc-L-lysine

storageNote1 : -30° to -10°C