Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nalpha-Benzoyl-L-ornithine, 98%

Click to view available options
Quantity:
250mg
1g
5g
17966-71-1
C12H16N2O3
236.271
MFCD00153452
PKONOCQSEOHHJW-JTQLQIEISA-N
n2-benzoyl-l-ornithine,bz-orn-oh,n-benzoyl-ornithine,s-5-amino-2-benzamidopentanoic acid,2s-5-amino-2-phenylformamido pentanoic acid,benzoylornithin,ambotzbaa0043,l-ornithine, n2-benzoyl,n-alpha-benzoyl-l-ornithine
7016391
(2S)-5-amino-2-benzamidopentanoic acid
C1=CC=C(C=C1)C(=O)NC(CCCN)C(=O)O
17966-71-1
C12H16N2O3
236.271
MFCD00153452
PKONOCQSEOHHJW-JTQLQIEISA-N
n2-benzoyl-l-ornithine,bz-orn-oh,n-benzoyl-ornithine,s-5-amino-2-benzamidopentanoic acid,2s-5-amino-2-phenylformamido pentanoic acid,benzoylornithin,ambotzbaa0043,l-ornithine, n2-benzoyl,n-alpha-benzoyl-l-ornithine
7016391
(2S)-5-amino-2-benzamidopentanoic acid
C1=CC=C(C=C1)C(=O)NC(CCCN)C(=O)O
Formula Weight 236.27
Physical Form Powder
Percent Purity 98%
Color White
Quantity 1g
Chemical Name or Material Nalpha-Benzoyl-L-ornithine

storageNote1 : -30° to -10°C