Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nalpha-Benzyloxycarbonyl-3-(Boc-amino)-D-alanine, 96%

Click to view available options
Quantity:
250mg
1g
5g
62234-36-0
C16H22N2O6
338.36
MFCD01318749
WJKGPJRAGHSOLM-GFCCVEGCSA-N
z-d-dap boc-oh,d-n-cbz-3-n-boc-amino-alanine,r-2-benzyloxy carbonyl amino-3-tert-butoxycarbonyl amino propanoic acid,n-a-z-n-beta-boc-d-2,3-diaminopropionic acid,n-alpha-benzyloxycarbonyl-n-beta-t-butyloxycarbonyl-d-2,3-diaminopropionic acid,r-2-benzyloxycarbonylamino-3-tert-butoxycarbonylamino-propionic acid,2r-2-benzyloxycarbonyl amino-3-tert-butoxycarbonyl amino propionic acid,3-1,1-dimethylethoxy carbonyl amino-n-phenylmethoxy carbonyl-d-alanine,2r-2-benzyloxy carbonyl amino-3-tert-butoxycarbonyl amino propanoic acid,2r-2-benzyloxy carbonyl amino-3-tert-butoxycarbonyl amino propanoic acid
7006678
(2R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-2-(phenylmethoxycarbonylamino)propanoic acid
CC(C)(C)OC(=O)NCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
62234-36-0
C16H22N2O6
338.36
MFCD01318749
WJKGPJRAGHSOLM-GFCCVEGCSA-N
z-d-dap boc-oh,d-n-cbz-3-n-boc-amino-alanine,r-2-benzyloxy carbonyl amino-3-tert-butoxycarbonyl amino propanoic acid,n-a-z-n-beta-boc-d-2,3-diaminopropionic acid,n-alpha-benzyloxycarbonyl-n-beta-t-butyloxycarbonyl-d-2,3-diaminopropionic acid,r-2-benzyloxycarbonylamino-3-tert-butoxycarbonylamino-propionic acid,2r-2-benzyloxycarbonyl amino-3-tert-butoxycarbonyl amino propionic acid,3-1,1-dimethylethoxy carbonyl amino-n-phenylmethoxy carbonyl-d-alanine,2r-2-benzyloxy carbonyl amino-3-tert-butoxycarbonyl amino propanoic acid,2r-2-benzyloxy carbonyl amino-3-tert-butoxycarbonyl amino propanoic acid
7006678
(2R)-3-[(2-methylpropan-2-yl)oxycarbonylamino]-2-(phenylmethoxycarbonylamino)propanoic acid
CC(C)(C)OC(=O)NCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
Formula Weight 338.36
Physical Form Solid
Percent Purity 96%
Quantity 250mg
Chemical Name or Material Nalpha-Benzyloxycarbonyl-3-(Boc-amino)-D-alanine

storageNote1 : -30° to -10°C