Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nalpha-Benzyloxycarbonyl-D-lysine, 95%

Click to view available options
Quantity:
1g
5g
70671-54-4
C14H20N2O4
280.324
MFCD00067713
OJTJKAUNOLVMDX-GFCCVEGCSA-N
z-d-lys-oh,cbz-d-lysine,nalpha-carbobenzoxy-d-lysine,nalpha-cbz-d-lysine,r-6-amino-2-benzyloxy carbonyl amino hexanoic acid,n-alpha-benzyloxycarbonyl-d-lysine,r-6-amino-2-benzyloxycarbonylamino hexanoic acid,n-cbz-d-lysine,nalpha-z-d-lysine,n,a-cbz-d-lysine
7017997
(2R)-6-amino-2-(phenylmethoxycarbonylamino)hexanoic acid
C1=CC=C(C=C1)COC(=O)NC(CCCCN)C(=O)O
70671-54-4
C14H20N2O4
280.324
MFCD00067713
OJTJKAUNOLVMDX-GFCCVEGCSA-N
z-d-lys-oh,cbz-d-lysine,nalpha-carbobenzoxy-d-lysine,nalpha-cbz-d-lysine,r-6-amino-2-benzyloxy carbonyl amino hexanoic acid,n-alpha-benzyloxycarbonyl-d-lysine,r-6-amino-2-benzyloxycarbonylamino hexanoic acid,n-cbz-d-lysine,nalpha-z-d-lysine,n,a-cbz-d-lysine
7017997
(2R)-6-amino-2-(phenylmethoxycarbonylamino)hexanoic acid
C1=CC=C(C=C1)COC(=O)NC(CCCCN)C(=O)O
Beilstein 5290234
Formula Weight 280.32
Physical Form Crystalline
Percent Purity 95%
Color White
Quantity 1g
Chemical Name or Material Nalpha-Benzyloxycarbonyl-D-lysine