Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nalpha-Benzyloxycarbonyl-Ndelta-Boc-L-ornithine, 98%

Click to view available options
Quantity:
5g
25g
7733-29-1
C18H26N2O6
366.414
MFCD00276410
RWQCKACYKKSOKK-CQSZACIVSA-N
z-d-orn boc-oh,nalpha-z-ndelta-boc-d-ornithine,n-cbz-n'-boc-l-ornithine,na-z-nd-boc-d-ornithine,n2-benzyloxy carbonyl-n5-tert-butoxycarbonyl-d-ornithine,n~2~-benzyloxy carbonyl-n~5~-tert-butoxycarbonyl-d-ornithine,2r-2-benzyloxy carbonyl amino-5-tert-butoxycarbonyl amino pentanoic acid,2r-5-2-methylpropan-2-yl oxycarbonylamino-2-phenylmethoxycarbonylamino pentanoic acid,r-2-benzyloxycarbonyl amino-5-tert-butyloxycarbonyl amino pentanoic acid
7350736
(2R)-5-[(2-methylpropan-2-yl)oxycarbonylamino]-2-(phenylmethoxycarbonylamino)pentanoic acid
CC(C)(C)OC(=O)NCCCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
7733-29-1
C18H26N2O6
366.414
MFCD00276410
RWQCKACYKKSOKK-CQSZACIVSA-N
z-d-orn boc-oh,nalpha-z-ndelta-boc-d-ornithine,n-cbz-n'-boc-l-ornithine,na-z-nd-boc-d-ornithine,n2-benzyloxy carbonyl-n5-tert-butoxycarbonyl-d-ornithine,n~2~-benzyloxy carbonyl-n~5~-tert-butoxycarbonyl-d-ornithine,2r-2-benzyloxy carbonyl amino-5-tert-butoxycarbonyl amino pentanoic acid,2r-5-2-methylpropan-2-yl oxycarbonylamino-2-phenylmethoxycarbonylamino pentanoic acid,r-2-benzyloxycarbonyl amino-5-tert-butyloxycarbonyl amino pentanoic acid
7350736
(2R)-5-[(2-methylpropan-2-yl)oxycarbonylamino]-2-(phenylmethoxycarbonylamino)pentanoic acid
CC(C)(C)OC(=O)NCCCC(C(=O)O)NC(=O)OCC1=CC=CC=C1
Beilstein 3086976
Formula Weight 366.41
Physical Form Solid
Percent Purity 98%
Melting Point 97°C to 103°C
Quantity 25g
Chemical Name or Material Nalpha-Benzyloxycarbonyl-Ndelta-Boc-L-ornithine