Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nalpha-Benzyloxycarbonyl-Nepsilon-Boc-L-lysine 4-nitrophenyl ester, 98%

Click to view available options
Quantity:
250mg
1g
5g
2212-69-3
C25H31N3O8
501.536
MFCD00076963
XBMRVOHLFIZDDV-NRFANRHFSA-N
z-lys boc-onp,s-4-nitrophenyl 2-benzyloxy carbonyl amino-6-tert-butoxycarbonyl amino hexanoate,4-nitrophenyl 2s-2-benzyloxy carbonyl amino-6-tert-butoxycarbonyl amino hexanoate,z-lys boc onp,z-n-epsilon-boc-l-lysine 4-nitrophenyl ester
13503265
(4-nitrophenyl) (2S)-6-[(2-methylpropan-2-yl)oxycarbonylamino]-2-(phenylmethoxycarbonylamino)hexanoate
CC(C)(C)OC(=O)NCCCCC(C(=O)OC1=CC=C(C=C1)[N+](=O)[O-])NC(=O)OCC2=CC=CC=C2
2212-69-3
C25H31N3O8
501.536
MFCD00076963
XBMRVOHLFIZDDV-NRFANRHFSA-N
z-lys boc-onp,s-4-nitrophenyl 2-benzyloxy carbonyl amino-6-tert-butoxycarbonyl amino hexanoate,4-nitrophenyl 2s-2-benzyloxy carbonyl amino-6-tert-butoxycarbonyl amino hexanoate,z-lys boc onp,z-n-epsilon-boc-l-lysine 4-nitrophenyl ester
13503265
(4-nitrophenyl) (2S)-6-[(2-methylpropan-2-yl)oxycarbonylamino]-2-(phenylmethoxycarbonylamino)hexanoate
CC(C)(C)OC(=O)NCCCCC(C(=O)OC1=CC=C(C=C1)[N+](=O)[O-])NC(=O)OCC2=CC=CC=C2
Formula Weight 501.54
Physical Form Solid
Percent Purity 98%
Quantity 250mg
Chemical Name or Material Nalpha-Benzyloxycarbonyl-Nepsilon-Boc-L-lysine 4-nitrophenyl ester

storageNote1 : 2° to 8°C