Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nalpha-Benzyloxycarbonyl-Nomega-(4-methoxybenzenesulfonyl)-L-arginine dicyclohexylammonium salt, 95%

Click to view available options
Quantity:
250mg
1g
5g
58810-11-0
C33H49N5O7S
659.843
MFCD00237326
YIFJUFKMGZAOPA-FERBBOLQSA-N
z-arg mbs-oh dcha,2s-2-benzyloxy carbonyl amino-5-n'-4-methoxybenzenesulfonyl carbamimidamido pentanoic acid; dicha,z-arg mbs-oh?dcha,e-n~5~-amino 4-methoxybenzene-1-sulfonyl amino methylidene-n~2~-benzyloxy carbonyl-l-ornithine-n-cyclohexylcyclohexanamine 1/1,n^a-benzyloxycarbonyl-n^w-4-methoxybenzenesulfonyl-l-arginine dicyclohexylammonium salt
56777357
(2S)-5-[[amino-[(4-methoxyphenyl)sulfonylamino]methylidene]amino]-2-(phenylmethoxycarbonylamino)pentanoic acid;N-cyclohexylcyclohexanamine
COC1=CC=C(C=C1)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OCC2=CC=CC=C2)N.C1CCC(CC1)NC2CCCCC2
58810-11-0
C33H49N5O7S
659.843
MFCD00237326
YIFJUFKMGZAOPA-FERBBOLQSA-N
z-arg mbs-oh dcha,2s-2-benzyloxy carbonyl amino-5-n'-4-methoxybenzenesulfonyl carbamimidamido pentanoic acid; dicha,z-arg mbs-oh?dcha,e-n~5~-amino 4-methoxybenzene-1-sulfonyl amino methylidene-n~2~-benzyloxy carbonyl-l-ornithine-n-cyclohexylcyclohexanamine 1/1,n^a-benzyloxycarbonyl-n^w-4-methoxybenzenesulfonyl-l-arginine dicyclohexylammonium salt
56777357
(2S)-5-[[amino-[(4-methoxyphenyl)sulfonylamino]methylidene]amino]-2-(phenylmethoxycarbonylamino)pentanoic acid;N-cyclohexylcyclohexanamine
COC1=CC=C(C=C1)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OCC2=CC=CC=C2)N.C1CCC(CC1)NC2CCCCC2
Formula Weight 659.84
Physical Form Solid
Percent Purity 95%
Quantity 250mg
Chemical Name or Material Nalpha-Benzyloxycarbonyl-Nomega-(4-methoxybenzenesulfonyl)-L-arginine dicyclohexylammonium salt

storageNote1 : -30° to -10°C