Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nalpha-Benzyloxycarbonyl-Nomega-nitro-L-arginine, 98%

Click to view available options
Quantity:
5g
25g
2304-98-5
C14H19N5O6
353.335
MFCD00038092
BZPCSFNCKORLQG-NSHDSACASA-N
z-arg no2-oh,z-l-arg no2-oh,n-cbz-n-nitro-l-arginine,cbz-l-arg no2-oh,s-2-benzyloxy carbonyl amino-5-3-nitroguanidino pentanoic acid,l-ornithine, n5-imino nitroamino methyl-n2-phenylmethoxy carbonyl,cbz-arg no2-oh,n5-imino nitroamino methyl-n2-phenylmethoxy carbonyl-l-ornithine,2s-2-benzyloxy carbonyl amino-5-n'-nitrocarbamimidamido pentanoic acid,n-alpha-z-n-omega-nitro-l-arginine
12695023
C1=CC=C(C=C1)COC(=O)NC(CCCN=C(N)N[N+](=O)[O-])C(=O)O
2304-98-5
C14H19N5O6
353.335
MFCD00038092
BZPCSFNCKORLQG-NSHDSACASA-N
z-arg no2-oh,z-l-arg no2-oh,n-cbz-n-nitro-l-arginine,cbz-l-arg no2-oh,s-2-benzyloxy carbonyl amino-5-3-nitroguanidino pentanoic acid,l-ornithine, n5-imino nitroamino methyl-n2-phenylmethoxy carbonyl,cbz-arg no2-oh,n5-imino nitroamino methyl-n2-phenylmethoxy carbonyl-l-ornithine,2s-2-benzyloxy carbonyl amino-5-n'-nitrocarbamimidamido pentanoic acid,n-alpha-z-n-omega-nitro-l-arginine
12695023
C1=CC=C(C=C1)COC(=O)NC(CCCN=C(N)N[N+](=O)[O-])C(=O)O
Formula Weight 353.33
Physical Form Solid
Percent Purity 98%
Quantity 5g
Chemical Name or Material Nalpha-Benzyloxycarbonyl-Nomega-nitro-L-arginine

EINECSNumber : 219-970-0

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold