Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nalpha-Boc-Ndelta-xanthyl-D-glutamine, 98%

Click to view available options
Quantity:
250mg
1g
5g
99092-88-3
C23H26N2O6
426.469
MFCD00151873
HOIIDGIJEPOSLL-MRXNPFEDSA-N
boc-d-gln xan-oh,ambotzbaa1180,n-alpha-boc-n-delta-xanthyl-d-glutamine,n-alpha-t-butyloxycarbonyl-n-gamma-xantyl-d-glutamine,n~2~-tert-butoxycarbonyl-n-9h-xanthen-9-yl-d-glutamine,2r-2-tert-butoxycarbonyl amino-4-9h-xanthen-9-yl carbamoyl butanoic acid
40424385
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxo-5-(9H-xanthen-9-ylamino)pentanoic acid
CC(C)(C)OC(=O)NC(CCC(=O)NC1C2=CC=CC=C2OC3=CC=CC=C13)C(=O)O
99092-88-3
C23H26N2O6
426.469
MFCD00151873
HOIIDGIJEPOSLL-MRXNPFEDSA-N
boc-d-gln xan-oh,ambotzbaa1180,n-alpha-boc-n-delta-xanthyl-d-glutamine,n-alpha-t-butyloxycarbonyl-n-gamma-xantyl-d-glutamine,n~2~-tert-butoxycarbonyl-n-9h-xanthen-9-yl-d-glutamine,2r-2-tert-butoxycarbonyl amino-4-9h-xanthen-9-yl carbamoyl butanoic acid
40424385
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxo-5-(9H-xanthen-9-ylamino)pentanoic acid
CC(C)(C)OC(=O)NC(CCC(=O)NC1C2=CC=CC=C2OC3=CC=CC=C13)C(=O)O
Formula Weight 426.47
Physical Form Powder
Percent Purity 98%
Color White
Quantity 5g
Chemical Name or Material Nalpha-Boc-Ndelta-xanthyl-D-glutamine

storageNote1 : -30° to -10°C