Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nalpha-Boc-Nepsilon-Fmoc-L-lysine methyl ester, 95%

Click to view available options
Quantity:
250mg
1g
133628-28-1
C27H34N2O6
482.577
MFCD00236856
NUSZDVABASFMMF-QHCPKHFHSA-N
boc-lys fmoc-ome,methyl 2s-2-tert-butoxycarbonyl amino-6-9h-fluoren-9-ylmethoxy carbonyl amino hexanoate,methyl n~2~-tert-butoxycarbonyl-n~6~-9h-fluoren-9-yl methoxy carbonyl-l-lysinate,l-lysine,n2-1,1-dimethylethoxy carbonyl-n6-9h-fluoren-9-ylmethoxy carbonyl-,methyl ester,methyl 2s-6-9h-fluoren-9-ylmethoxycarbonylamino-2-2-methylpropan-2-yl oxycarbonylamino hexanoate
56776966
methyl (2S)-6-(9H-fluoren-9-ylmethoxycarbonylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoate
CC(C)(C)OC(=O)NC(CCCCNC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)C(=O)OC
133628-28-1
C27H34N2O6
482.577
MFCD00236856
NUSZDVABASFMMF-QHCPKHFHSA-N
boc-lys fmoc-ome,methyl 2s-2-tert-butoxycarbonyl amino-6-9h-fluoren-9-ylmethoxy carbonyl amino hexanoate,methyl n~2~-tert-butoxycarbonyl-n~6~-9h-fluoren-9-yl methoxy carbonyl-l-lysinate,l-lysine,n2-1,1-dimethylethoxy carbonyl-n6-9h-fluoren-9-ylmethoxy carbonyl-,methyl ester,methyl 2s-6-9h-fluoren-9-ylmethoxycarbonylamino-2-2-methylpropan-2-yl oxycarbonylamino hexanoate
56776966
methyl (2S)-6-(9H-fluoren-9-ylmethoxycarbonylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoate
CC(C)(C)OC(=O)NC(CCCCNC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)C(=O)OC
Applications
It is employed as a intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in freezer -20°C. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Formula Weight 482.58
Physical Form Powder
Percent Purity 95%
Color White
Quantity 1g
Chemical Name or Material Nalpha-Boc-Nepsilon-Fmoc-L-lysine methyl ester

storageNote1 : -30° to -10°C