Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nalpha-Boc-Nepsilon-trifluoroacetyl-D-lysine, 95%

Click to view available options
Quantity:
250mg
1g
96561-04-5
C13H21F3N2O5
342.315
MFCD00236857
DEIYNDIFGSDDCY-MRVPVSSYSA-N
boc-d-lys tfa-oh,r-2-tert-butoxycarbonyl amino-6-2,2,2-trifluoroacetamido hexanoic acid,2r-2-tert-butoxycarbonyl amino-6-2,2,2-trifluoroacetamido hexanoic acid,boc-d-lys tfa,nalpha-boc-nepsilon-trifluoroacetyl-d-lysine,d-lysine,n2-1,1-dimethylethoxy carbonyl-n6-trifluoroacetyl-9ci,r-2-tert-butoxycarbonyl amino-6-2,2,2-trifluoroacetamido hexanoicacid,2r-2-2-methylpropan-2-yl oxycarbonylamino-6-2,2,2-trifluoroacetyl amino hexanoic acid
6993446
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-6-[(2,2,2-trifluoroacetyl)amino]hexanoic acid
CC(C)(C)OC(=O)NC(CCCCNC(=O)C(F)(F)F)C(=O)O
96561-04-5
C13H21F3N2O5
342.315
MFCD00236857
DEIYNDIFGSDDCY-MRVPVSSYSA-N
boc-d-lys tfa-oh,r-2-tert-butoxycarbonyl amino-6-2,2,2-trifluoroacetamido hexanoic acid,2r-2-tert-butoxycarbonyl amino-6-2,2,2-trifluoroacetamido hexanoic acid,boc-d-lys tfa,nalpha-boc-nepsilon-trifluoroacetyl-d-lysine,d-lysine,n2-1,1-dimethylethoxy carbonyl-n6-trifluoroacetyl-9ci,r-2-tert-butoxycarbonyl amino-6-2,2,2-trifluoroacetamido hexanoicacid,2r-2-2-methylpropan-2-yl oxycarbonylamino-6-2,2,2-trifluoroacetyl amino hexanoic acid
6993446
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-6-[(2,2,2-trifluoroacetyl)amino]hexanoic acid
CC(C)(C)OC(=O)NC(CCCCNC(=O)C(F)(F)F)C(=O)O
Formula Weight 342.32
Physical Form Solid
Percent Purity 95%
Quantity 1g
Chemical Name or Material Nalpha-Boc-Nepsilon-trifluoroacetyl-D-lysine

storageNote1 : -30° to -10°C