Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nalpha-Fmoc-3-(Fmoc-amino)-L-alanine, 95%

Click to view available options
Quantity:
250mg
1g
5g
201473-90-7
C33H28N2O6
548.595
MFCD00237005
CEEGOSWFFHSPHM-PMERELPUSA-N
fmoc-dap fmoc-oh,s-2,3-bis 9h-fluoren-9-yl methoxy carbonyl amino propanoic acid,2s-2,3-bis 9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid,ambotzfaa1717,fmoc-dap fmoc-oh hplc,n-a,n-b-di-fmoc-l-2,3-diaminopropionic acid,2s-2,3-bis 9h-fluoren-9-ylmethoxycarbonylamino propanoic acid,n-alpha-n-beta-bis-9-fluorenylmethyloxycarbonyl-l-2,3-diaminopropionic acid
2756104
(2S)-2,3-bis(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46
201473-90-7
C33H28N2O6
548.595
MFCD00237005
CEEGOSWFFHSPHM-PMERELPUSA-N
fmoc-dap fmoc-oh,s-2,3-bis 9h-fluoren-9-yl methoxy carbonyl amino propanoic acid,2s-2,3-bis 9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid,ambotzfaa1717,fmoc-dap fmoc-oh hplc,n-a,n-b-di-fmoc-l-2,3-diaminopropionic acid,2s-2,3-bis 9h-fluoren-9-ylmethoxycarbonylamino propanoic acid,n-alpha-n-beta-bis-9-fluorenylmethyloxycarbonyl-l-2,3-diaminopropionic acid
2756104
(2S)-2,3-bis(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCC(C(=O)O)NC(=O)OCC4C5=CC=CC=C5C6=CC=CC=C46
Formula Weight 548.6
Physical Form Crystalline
Percent Purity 95%
Color White
Quantity 250mg
Chemical Name or Material Nalpha-Fmoc-3-(Fmoc-amino)-L-alanine

storageNote1 : 2° to 8°C