Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nalpha-Fmoc-L-histidine, 98%

Click to view available options
Quantity:
5g
25g
116611-64-4
C21H19N3O4
377.4
MFCD00190885
SIRPVCUJLVXZPW-IBGZPJMESA-N
fmoc-l-histidine,fmoc-his-oh,n-fmoc-l-histidine,fmoc-his,nalpha-fmoc-l-histidine,s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-1h-imidazol-4-yl propanoic acid,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-1h-imidazol-4-yl propanoic acid,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-1h-imidazol-4-yl propanoic acid,pubchem18623,fmoc-l-his-oh
7019075
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(1H-imidazol-5-yl)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CN=CN4)C(=O)O
116611-64-4
C21H19N3O4
377.4
MFCD00190885
SIRPVCUJLVXZPW-IBGZPJMESA-N
fmoc-l-histidine,fmoc-his-oh,n-fmoc-l-histidine,fmoc-his,nalpha-fmoc-l-histidine,s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-1h-imidazol-4-yl propanoic acid,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-1h-imidazol-4-yl propanoic acid,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-1h-imidazol-4-yl propanoic acid,pubchem18623,fmoc-l-his-oh
7019075
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(1H-imidazol-5-yl)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CN=CN4)C(=O)O
Formula Weight 377.4
Physical Form Solid
Percent Purity 98%
Quantity 25g
Chemical Name or Material Nalpha-Fmoc-L-histidine

storageNote1 : -30° to -10°C