Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nalpha-Fmoc-Nepsilon-carbamoyl-L-lysine, 95%

Click to view available options
Quantity:
250mg
1g
5g
201485-17-8
C22H25N3O5
411.458
MFCD00237387
VJZUCXPETVPQIB-IBGZPJMESA-N
fmoc-hocit-oh,fmoc-l-homocitrulline,fmoc-hcit-oh,s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-ureidohexanoic acid,fmoc-homocit-oh,2s-6-carbamoylamino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid,ambotzfaa1383,fmoc-lys carbamoyl-oh,n-alpha-9-fluorenylmethyloxycarbonyl-l-homocitrulline,s-2-9h-fluoren-9-yl methoxy carbonyl-amino-6-ureidohexanoic acid
7019813
(2S)-6-(carbamoylamino)-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCCCNC(=O)N)C(=O)O
201485-17-8
C22H25N3O5
411.458
MFCD00237387
VJZUCXPETVPQIB-IBGZPJMESA-N
fmoc-hocit-oh,fmoc-l-homocitrulline,fmoc-hcit-oh,s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-ureidohexanoic acid,fmoc-homocit-oh,2s-6-carbamoylamino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid,ambotzfaa1383,fmoc-lys carbamoyl-oh,n-alpha-9-fluorenylmethyloxycarbonyl-l-homocitrulline,s-2-9h-fluoren-9-yl methoxy carbonyl-amino-6-ureidohexanoic acid
7019813
(2S)-6-(carbamoylamino)-2-(9H-fluoren-9-ylmethoxycarbonylamino)hexanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCCCNC(=O)N)C(=O)O
Formula Weight 411.46
Physical Form Solid
Percent Purity 95%
Quantity 250mg
Chemical Name or Material Nalpha-Fmoc-Nepsilon-carbamoyl-L-lysine

storageNote1 : -30° to -10°C