Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nalpha-Fmoc-Nomega-nitro-L-arginine, 95%

Click to view available options
Quantity:
5g
25g
58111-94-7
C21H23N5O6
441.444
MFCD00065617
RXMHIKWOZKQXCJ-SFHVURJKSA-N
fmoc-arg no2-oh,n-fmoc-n'-nitro-l-arginine,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-n'-nitrocarbamimidamido pentanoic acid,fmoc-arg no-oh,pubchem18946,fmoc-arg no2-oh,fmoc-nw-nitro-l-arginine,n alpha-fmoc-n omega-nitro-l-arginine,fmoc-arg no2-oh hplc,n-alpha-9-fluorenylmethyloxycarbonyl-n'-nitro-l-arginine
12096824
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCCN=C(N)N[N+](=O)[O-])C(=O)O
58111-94-7
C21H23N5O6
441.444
MFCD00065617
RXMHIKWOZKQXCJ-SFHVURJKSA-N
fmoc-arg no2-oh,n-fmoc-n'-nitro-l-arginine,2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-5-n'-nitrocarbamimidamido pentanoic acid,fmoc-arg no-oh,pubchem18946,fmoc-arg no2-oh,fmoc-nw-nitro-l-arginine,n alpha-fmoc-n omega-nitro-l-arginine,fmoc-arg no2-oh hplc,n-alpha-9-fluorenylmethyloxycarbonyl-n'-nitro-l-arginine
12096824
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCCN=C(N)N[N+](=O)[O-])C(=O)O
Beilstein 7060334
Formula Weight 441.44
Physical Form Powder
Percent Purity 95%
Color White
Quantity 25g
Chemical Name or Material Nalpha-Fmoc-Nomega-nitro-L-arginine

storageNote1 : 2° to 8°C