Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Nalpha-Fmoc-Nomega-(p-toluenesulfonyl)-L-arginine, 95%

Click to view available options
Quantity:
5g
25g
100g
83792-47-6
C28H30N4O6S
550.63
MFCD00063331
JRRARHJPRLAGNT-VWLOTQADSA-N
fmoc-arg tos-oh,nalpha-fmoc-nomega-tosyl-l-arginine,nalpha-9h-fluoren-9-ylmethoxy carbonyl-nomega-tosyl-l-arginine,fmoc-arginine tos,fmoc-l-arg tos-oh,arginine derivative 6,pubchem18947,fmoc-arg tos-oh et2o,fmoc-n-gamma-tosyl-l-arginine etherate,fmoc-nw-4-toluenesulfonyl-l-arginine
3010412
(2S)-5-[[amino-[(4-methylphenyl)sulfonylamino]methylidene]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid
CC1=CC=C(C=C1)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)N
83792-47-6
C28H30N4O6S
550.63
MFCD00063331
JRRARHJPRLAGNT-VWLOTQADSA-N
fmoc-arg tos-oh,nalpha-fmoc-nomega-tosyl-l-arginine,nalpha-9h-fluoren-9-ylmethoxy carbonyl-nomega-tosyl-l-arginine,fmoc-arginine tos,fmoc-l-arg tos-oh,arginine derivative 6,pubchem18947,fmoc-arg tos-oh et2o,fmoc-n-gamma-tosyl-l-arginine etherate,fmoc-nw-4-toluenesulfonyl-l-arginine
3010412
(2S)-5-[[amino-[(4-methylphenyl)sulfonylamino]methylidene]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid
CC1=CC=C(C=C1)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)N
Beilstein 5198802
Formula Weight 550.63
Physical Form Powder
Percent Purity 95%
Color White
Melting Point 80°C
Quantity 100g
Chemical Name or Material Nalpha-Fmoc-Nomega-(p-toluenesulfonyl)-L-arginine

storageNote1 : 2° to 8°C