Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha-Fmoc-Nomega-(p-toluenesulfonyl)-L-arginine, 95%, Thermo Scientific™

Catalog Number AAH6635106
Click to view available options
Quantity:
5 g
25 g
100 g
83792-47-6
C28H30N4O6S
550.63
MFCD00063331
JRRARHJPRLAGNT-VWLOTQADSA-N
fmoc-arg tos-oh, nalpha-fmoc-nomega-tosyl-l-arginine, nalpha-9h-fluoren-9-ylmethoxy carbonyl-nomega-tosyl-l-arginine, fmoc-arginine tos, fmoc-l-arg tos-oh, arginine derivative 6, pubchem18947, fmoc-arg tos-oh et2o, fmoc-n-gamma-tosyl-l-arginine etherate, fmoc-nw-4-toluenesulfonyl-l-arginine
3010412
(2S)-5-[[amino-[(4-methylphenyl)sulfonylamino]methylidene]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid
CC1=CC=C(C=C1)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)N
83792-47-6
C28H30N4O6S
550.63
MFCD00063331
JRRARHJPRLAGNT-VWLOTQADSA-N
fmoc-arg tos-oh, nalpha-fmoc-nomega-tosyl-l-arginine, nalpha-9h-fluoren-9-ylmethoxy carbonyl-nomega-tosyl-l-arginine, fmoc-arginine tos, fmoc-l-arg tos-oh, arginine derivative 6, pubchem18947, fmoc-arg tos-oh et2o, fmoc-n-gamma-tosyl-l-arginine etherate, fmoc-nw-4-toluenesulfonyl-l-arginine
3010412
(2S)-5-[[amino-[(4-methylphenyl)sulfonylamino]methylidene]amino]-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid
CC1=CC=C(C=C1)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 80°C
Color White
Beilstein 5198802
Formula Weight 550.63
Percent Purity 95%
Quantity 5 g
Physical Form Powder
Chemical Name or Material Nalpha-Fmoc-Nomega-(p-toluenesulfonyl)-L-arginine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit