Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nalpha,Nepsilon-Bis(2,4-dinitrophenyl)-L-lysine 97.0+%, TCI America™
SDP

Catalog No. D1051100MG
Click to view available options
Quantity:
100 mg

Chemical Identifiers

CAS 1655-49-8
Molecular Formula C18H18N6O10
Molecular Weight (g/mol) 478.374
MDL Number MFCD00056105
InChI Key XFVSVNWYIWINEC-HNNXBMFYSA-N
Synonym Nalpha,Nepsilon-Di-Dnp-L-lysine, Dnp-Lys(Dnp)-OH
PubChem CID 96818
IUPAC Name (2S)-2,6-bis(2,4-dinitroanilino)hexanoic acid
SMILES C1=CC(=C(C=C1[N+](=O)[O-])[N+](=O)[O-])NCCCCC(C(=O)O)NC2=C(C=C(C=C2)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point 176°C
Color Yellow
Quantity 100 mg
UN Number 1325
Formula Weight 478.37
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha,Nepsilon-Bis(2,4-dinitrophenyl)-L-lysine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.