Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Naltriben mesylate, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
122517-78-6
C27H29NO7S
511.589
XRRFZOCDAWPIBB-UHFFFAOYSA-N
naltriben mesylate
71296991
CS(=O)(=O)O.C1CC1CN2CCC34C5C6=C(CC3(C2CC7=C4C(=C(C=C7)O)O5)O)C8=CC=CC=C8O6
122517-78-6
C27H29NO7S
511.589
XRRFZOCDAWPIBB-UHFFFAOYSA-N
naltriben mesylate
71296991
CS(=O)(=O)O.C1CC1CN2CCC34C5C6=C(CC3(C2CC7=C4C(=C(C=C7)O)O5)O)C8=CC=CC=C8O6

Standard δ2 selective antagonist

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Assay Percent Range >98%
Formula Weight 511.59
Chemical Name or Material Naltriben mesylate

Recommended Storage : Desiccate at Room Temperature