Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Naltriben mesylate, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Selective and potent δ-opioid receptor antagonist (Ki values are 0.013, 19 and 152 nM for δ, μ and κ receptors respectively). Displays selectivity for the δ2 subtype in vivo.
Specifications
Quantity | 50 mg |
Formula Weight | 511.59 |
Percent Purity | >98% |
Chemical Name or Material | Naltriben mesylate |
Safety and Handling
Recommended Storage | Desiccate at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction