Learn More
NAN-190 hydrobromide, Tocris
5-HT1A antagonist
Manufacturer:  R&D Systems 0553/250
Quantity 50mg
 View more versions of this product
Catalog No. 05-532-50
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>98%
474.4
NAN-190 hydrobromide
C23H27N3O3·HBr
1-(2-Methoxyphenyl)-4-(4-phthalimidobutyl)piperazine hydrobromide
Specifications
115338-32-4
C23H27N3O3·HBr
50mg
NAN-190 hydrobromide
>98%
474.4
1-(2-Methoxyphenyl)-4-(4-phthalimidobutyl)piperazine hydrobromide
Safety and Handling
Recommended Storage : Store at Room Temperature