Learn More
Selleck Chemical LLCSupplier Diversity Partner Nanchangmycin 50mg 65101-87-3  DFS Item
Manufacturer:  Selleck Chemical LLCSupplier Diversity Partner S145050MG
Nanchangmycin is a polyether antibiotic with similar structure to dianemycin and is very active against a broad spectrum of harmful nematodes and insects but not for mammals and plants.
Catalog No. 50-719-9