Learn More
Promega NanoBRET™ Nano-Glo™ Substrate
The NanoBRET™ Nano-Glo(R) Substrate is the substrate used by NanoLuc(R) Luciferase to generate the donor signal in several NanoBRET™ Assays.
$396.50 - $10940.00
Specifications
Content And Storage -30°C to -10°C
View More Specs
Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PRN1572
View Documents
Promega
N1572
5 x 50 μL
Each for $1,344.00
Add to cart
PRN1571
View Documents
Promega
N1571
50 μL
Each for $396.50
Add to cart
PRN1573
View Documents
Promega
N1573
2 x 1.25 mL
Each for $10,940.00
Add to cart
Description
Description
For use with NanoBRET assay, a proximity-based assay that can detect protein interactions by measuring energy transfer from a bioluminescent protein donor to a fluorescent protein acceptor.
Specifications
Specifications
-30°C to -10°C