Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ NanoDrop™ 2000/2000c Spectrophotometers

New sales of this product have been discontinued. Please see the newest model: Thermo Scientific NanoDrop One Microvolume UV-Vis Spectrophotometer.

Supplier:  Thermo Scientific™ ND2000CUSCAN

Request Bulk Order

 View more versions of this product

Catalog No. ND2000C

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Explore available promotions

Description

Description

New sales of this product have been discontinued. Please see the newest model: Thermo Scientific NanoDrop One Microvolume UV-Vis Spectrophotometer.

The patented sample retention system of the NanoDrop 2000 and 2000c spectrophotometers allows a sample to be pipetted directly onto an optical measurement surface. Once the measurement is complete, the surfaces are simply wiped with a lint-free lab wipe. The NanoDrop 2000c integrates the sample retention system with cuvette capability.

NanoDrop 2000

  • Micro-volume sample size (0.5-2.0μL)
  • Easy to use—pipet sample directly onto the pedestal
  • No dilutions required, even for highly concentrated samples
  • Fast measurement time of less than 5 seconds
  • Full spectral output (190 - 840 nm)
  • Preconfigured methods for A260, A280, labeled DNA or Proteins, and much more
  • User-friendly software that includes custom methods and data export capabilities

NanoDrop 2000c

  • Combines all features of the NanoDrop 2000 with cuvette capability in one instrument
  • Advanced micro-volume pedestal technology
  • Dual modes—choose cuvette or pedestal
  • Broader concentration range for measuring very low or very high concentrations
  • Cuvette capability allows for kinetics (time or time/temperature studies) and cell culture (OD 600) measurements

NanoDrop 2000c - Cuvette

  • Heating: 37°C, ±0.5°C
  • Stirring: 150 - 850rpm
  • Z-Height: 8.5mm
  • Cuvette dimensions: 12.5 x 12.5mm, up to 48mmH
  • Cuvette path length options: 10, 5, 2 and 1mm
  • Type: Masked cuvette
  • Absorbance Range: 0.008 to 1.5
  • Measurement Time: <3 sec.
  • Weight: 2.1kg

Compatible with:

Professional versions of Microsoft™ Windows™ XP (32-bit) w/Service Pack (SP) 2 or later, Vista™ (32-bit), and Windows™ 7 (32 bit and 64 bit)

TRUSTED_SUSTAINABILITY
Specifications

Specifications

3% (at 0.74 Abs at 350 nm)
0.002
Microsoft Windows 7 Professional (32-bit and 64-bit), Windows 8 (32- and 64-bit), and Windows 10 Pro (64-bit)
20 cm
Pedestal: 2 ng/μL (dsDNA), 0.10 mg/mL (BSA) Cuvette: 0.4 ng/μL (dsDNA), 0.01 mg/mL (BSA)
2048-element Linear Silicon CCD Array
Spectrophotometer
190 to 840 nm
Nucleic Acid Quantification, DNA Quantification, RNA Quantification, Protein Quantification
<3 sec.
1
10, 5, 2, 1 mm
1000, 0.5 to 2.0 μL
Microsoft Windows 7 Professional (32-bit and 64-bit), Windows 8 (32- and 64-bit), and Windows 10 Pro (64-bit)
1 year parts and labor
±1 nm
14 cm
Pedestal: 0 to 300 A Cuvette: 0 to 1.5 A
UL/CSA and CE
USB
Microvolume Spectrophotometer, with cuvet capability
2 to 15,000n g/μL (dsDNA) , 0.10 to 400 mg/mL (BSA)
Xenon Flash
12 VDC
14 x 20 cm
n.a.
NanoDrop 2000c
37°; ±0.5°C
12 V DC
1.8 nm (FWHM at Hg 253.7)
0.5 to 2.0 μL
5 W
2.0 kg
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Accessories
Promotions

Promotions

Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

US Patents 6,628,382 and 6,809,826.