Learn More
Thermo Scientific™ NanoDrop™ 3300 Fluorospectrometer Available on GSA/VA Contract for Federal Government customers only.
Choose this versatile, cost-effective fluorospectrometer for broad spectrum fluorescence analysis.
$13900.00 - $15900.00
Specifications
Certifications/Compliance UL/CSA and CE
Connections USB
Detectors 2048-element linear silicon CCD array
Function Fluorescence spectroscopy
Type Fluorospectrometer
View More Specs
Products 2
Catalog Number Mfr. No. Model Price Quantity & Availability  
Catalog Number Mfr. No. Model Price Quantity & Availability  
ND3300 Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Thermo Scientific™
ND3300
NanoDrop 3300 Fluorospectrometer Each for $13,900.00
Add to cart
13-400-414 Available on GSA/VA Contract for Federal Government customers only.
Accessories View Documents
Thermo Scientific™
ND3300LAPTOP
NanoDrop 3300 Fluorospectrometer with Laptop Computer Each for $15,900.00
Add to cart
Description
Description
Choose the versatile, cost-effective Thermo Scientific™ NanoDrop 3300 Fluorospectrometer for broad spectrum fluorescence analysis. Using a patented technology, it uses as little as 1 µL of sample volume.
The NanoDrop 3300 significantly lowers the mass detection limit more than an order of magnitude, compared to conventional fluorometers. With its advanced features and analysis options, the NanoDrop 3300 allows for a diverse range of measurements from routine RiboGreen, PicoGreen, and GFP to advanced nanoparticle and FRET analysis.
  • Small footprint, fast measurement, and easy to use
  • Sample size as small as 1µL
  • UV, Blue and White LED excitation, covering a broad wavelength range
  • No filter changes
  • No expensive monochromator
  • Ability to run diverse applications
  • Customize applications specific to your research
  • Measures multiple fluorophores from a single sample
  • Tools to eliminate the guesswork
  • Provides both spectral and quantitative data
  • No external power supply is required; all operating power is supplied by the USB port on the PC.
Compatible with:
Professional versions of Microsoft™ Windows™ 7 (32 and 64 bit), Professional version of Windows 8.1 (64 bit), Windows 10 Pro (64 bit)
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
UL/CSA and CE
2048-element linear silicon CCD array
Fluorospectrometer
USB
Fluorescence spectroscopy
1µL
US Patents 6,628,382 and 6,809,826.