Thermo Scientific™ NanoDrop™ 8000 Spectrophotometer Available on GSA/VA Contract for Federal Government customers only.

Thermo Scientific™ NanoDrop 8000 is a muti-sample micro-volume UV-Vis spectrophotometer.

Manufacturer: Thermo Scientific™ ND8000LAPTOP

 View more versions of this product

Catalog No. 13-400-413

  Check Availability
Add to Cart

Disclaimers:

US Patents 6,628,382 and 6,809,826.


    Specifications

    Cycle Time <20 sec. (for 8 samples)
    Measurement Time <20 sec.
    Warranty One year parts and labor
    Compatibility Microsoft™ Windows™ XP (32-bit) w/Service Pack (SP) 2 or later, Vista™ (32-bit), and Windows 7 (32 bit and 64 bit)
    Connections USB
    Depth (Metric) 32cm
    Detection Range 2.5-3,700ng/µL (dsDNA), 0.15 - 100mg/mL (BSA)
    Detector Type 2048-element linear silicon CCD array
    Footprint 24 x 32 cm
    Accuracy (Absorption) 3%
    Sample Volume (Metric) 1µL
    Spectral Resolution ≤3nm (FWHM at Hg 546.1 nm)
    System Requirements Microsoft Windows XP (32-bit) w/Service Pack (SP) 2 or later,Vista (32-bit), and Windows 7 (32 bit and 64 bit)
    Type Spectrophotometer
    Volume (Metric) 1µL
    Voltage 12VDC
    Wattage ±1 nm
    Wavelength Range 220 - 750 nm
    Certifications/Compliance UL/CSA and CE
    Description Microvolume spectrometer with laptop
    Detection Limits 2.5ng/µL (dsDNA)
    Lamp Xenon flash lamp
    No. of Samples 8
    Pathlength (Metric) 0.1, 0.2 and 1.0 mm
    Wavelength Accuracy 1nm
    Absorbance Accuracy 3% (at 0.74 Abs at 350 nm)
    Absorbance Range 0 - 75Abs
    Accuracy 0.003 *SD of 10 individual measurements at 0.74Abs
    Power Consumption 30w
    Applications Nucleic acid, protein, cell culture and custom methods

    Thermo Scientific™ NanoDrop 8000 is a muti-sample micro-volume UV-Vis spectrophotometer. With the NanoDrop 8000, you can measure more samples in less time with the same reliability and patented sample retention technology of the NanoDrop 2000.

    The NanoDrop 8000 allows scientists to quickly and easily quantify and assess purity of samples such as proteins and nucleic acids. Using an 8-channel pipetter to dispense samples onto an array of pedestals, analyze 96 samples in less than six minutes.

    Key Features and Benefits

    • Designed for micro-volume (1µL) samples
    • No dilutions required, even for high concentration samples
    • Direct, easy measurements from tubes or 96-well plates
    • Can measure multiple samples in one measurement cycle, up to eight samples at one time
    • Single sample mode
    • Preconfigured methods for common applications such as Nucleic Acid, Protein A280, Microarray, Protein and Labels, Pierce 660, Bradford, BCA, Lowry and more
    • User-friendly software for creating custom methods and options to design reports and export data
    • Patented sample retention system eliminates the need for cuvets
    • Full spectrum (220 to 750nm)
    • Calculates sample purity ratios (260/280)

    Includes:

    USB cable and operating software

    Compatible with:

    Microsoft™ Windows™ XP (32-bit) w/Service Pack (SP) 2 or later, and Vista™ (32-bit).

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit