Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ NanoDrop™ 8000 Spectrophotometer PROMO

Thermo Scientific™ NanoDrop™ 8000 Spectrophotometer is a muti-sample, micro-volume, UV-Vis spectrophotometer.

Specifications

Absorbance Accuracy 3% (at 0.74 Abs at 350 nm)
Absorbance Range 0 to 75 Abs
Accuracy 0.003 SD of 10 individual measurements at 0.74 Abs
Certifications/Compliance UL/CSA and CE
Compatibility Professional versions of Microsoft™ Windows™ 7 (32 bit and 64 bit), Windows Vista (32 bit), and Windows XP (32 bit).
View More Specs

Products 1
Catalog Number Mfr. No. Description Price Quantity & Availability  
Catalog Number Mfr. No. Description Price Quantity & Availability  
ND8000PMO
View Documents
Thermo Scientific™
840281300PRM19
Basic Unit This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Allows scientists to quickly and easily quantify and assess purity of samples such as proteins and nucleic acids. Using an 8-channel pipetter to dispense samples onto an array of pedestals, analyze 96 samples in less than six minutes.

Key Features and Benefits

  • Designed for micro-volume (1 µL) samples
  • No dilutions required, even for high concentration samples
  • Direct, easy measurements from tubes or 96-well plates
  • Can measure multiple samples in one measurement cycle, up to eight samples at one time
  • Single sample mode
  • Preconfigured methods for common applications such as Nucleic Acid, Protein A280, Microarray, Protein and Labels, Pierce 660, Bradford, BCA, Lowry and more
  • User-friendly software for creating custom methods and options to design reports and export data
  • Patented sample retention system eliminates the need for cuvets
  • Full spectrum (220 to 750 nm)
  • Calculates sample purity ratios (260/280)

Includes:

USB cable and operating software

Compatible with:

Microsoft Windows 7 Professional (32-bit and 64-bit), Windows 8 (32- and 64-bit), and Windows 10 Pro (64-bit)

Specifications

Specifications

3% (at 0.74 Abs at 350 nm)
0.003 SD of 10 individual measurements at 0.74 Abs
Professional versions of Microsoft™ Windows™ 7 (32 bit and 64 bit), Windows Vista (32 bit), and Windows XP (32 bit).
2.5 ng/μL (dsDNA)
2048-element linear silicon CCD array
Xenon Flash Lamp
24 x 32 cm
Up to 8
≤3 nm (FWHM at Hg 546.1 nm)
1 year parts and labor
0 to 75 Abs
UL/CSA and CE
USB
2.5 to 3700 ng/μL (dsDNA), 0.15 to 100 mg/mL (BSA)
Nucleic acid, protein, cell culture, and custom methods
1 μL
<20 sec.
0.2 and 1.0 mm
Spectrophotometer
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit