Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ NanoDrop™ One Microvolume UV-Vis Spectrophotometer PROMO

Maximize your productivity with the Thermo Scientific™ NanoDrop™ One Spectrophotometer to quantify and qualify samples before your next experiment.

Manufacturer:  Thermo Scientific™ 840274100PR2

Catalog No. 13-400-518PR2

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Product Features:

  • Full spectrum microvolume UV-Vis spectrophotometer with 190nm-850nm wavelength measurement range
  • Sample vol 1.0 - 2.0µL
  • Measuremetn range 2 - 27500ng/µL dsDNA
  • Column integrity sensor
  • Tilting and sliding screen
  • 2 yr warranty

Quantify and qualify DNA, RNA, and protein samples with only 1-2µL and have full-spectral data in seconds using the Thermo Scientific NanoDrop One Microvolume UV-Vis spectrophotometer.

  • Prevent costly delays — Identify contaminants and obtain corrected concentrations
  • Collect data fast — Auto-Blank and Auto-Measure modes save time while organizing data
  • Save bench space — User-friendly local control with sleek, standalone design means no PC required

Thermo Scientific Acclaro Sample Intelligence Software enhances user understanding of sample quality while delivering accurate quantitative measurements. Early identification of contaminants prevent failure of downstream applications and save days of troubleshooting.

  • Verify sample quality in seconds — Identify sample contaminants and obtain corrected concentration results
  • Access information through alerts — Receive instant feedback about sample quality with on-demand technical support and guided troubleshooting
  • Rely on accurate results — Ensure measurement integrity with embedded sensor and digital image analysis

NanoDrop One Enhancements

  • Ergonomic, standalone design — Integrated Android tablet eliminates the need for a separate computer and provides modern connectivity for seamless data transfer via Wireless*, Ethernet or USB.
  • Auto-Measure and Auto-Blank functions — Multi-sample processing can be streamlined with instant measurements that occur when the pedestal arm is down. These functions can be turned ON or OFF with a single tap on the screen.
  • Wider dynamic range — No dilutions required for highly concentrated samples (dsDNA at 27,500 ug/µL) with auto-range pathlength technology.
  • Integrated Learning Center — Archive of answers with technical support documents and educational animations


NanoDrop One/Onec Spectrophotometer capabilities:

  • Wide spectral range (190 to 850 nm) for measuring a variety of samples types:
    • Peptides (205 nm)
    • DNA and RNA (260 nm)
    • Purified protein (280 nm)
    • Toxicology assays and industrial dyes (490 nm)
    • Gold nanoparticles (520 nm)
    • Colorimetric protein assays (BCA 562 nm, Bradford 595 nm, Modified Lowry 650 nm, Pierce 660 660 nm)
    • Optical Density measurements (600 nm)
  • Combining the patented** sample-retention system with cuvette capability accommodates low and high concentrations (2.0 - 27,500ng/µL dsDNA and 0.06 - 820mg/mL BSA)
  • Pedestal measurements that require only 0.5 to 2µL of your sample and do not require sample dilution, even for highly concentrated samples
  • Pre-configured methods for DNA, Protein A280, Microarray, Protein and Labels, Pierce 660, Bradford, BCA, and Lowry
  • User-friendly software that includes custom methods and data export capabilities
  • Instrument comes with a 2-year warranty for worry-free support


* Wi-Fi model not available in certain countries - consult a NanoDrop distributor
**Patents US6628382 and US6809826
Android is a trademark of Google Inc.

For research use only. Not for diagnostic purposes.
All NanoDrop instruments are approved to CE and UL/CSA.

Specifications

Specifications

PR-1 pedestal conditioning solution, PV-1 performance verification solution, USB memory device, Microfiber screen wipe, USA Power Cable
2 year
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit