Learn More
Waters Corp Waters nanoEase M/Z Peptide CSH C18 Column, 130Å, 1.7 μm, 300 μm X 100 mm
Supplier:  Waters Corp 186009255
Encompass_Preferred
Engineered for optimal performance in nano- to micro-scale liquid chromatographic separation, these columns provide superior sensitivity, resolution, and reproducibility for biomarker discovery and protein identification. They feature an ultra-low dispersion design and patented CSH Technology with 130Å pore, C18, 1.7 μm particles, offering excellent peak shape for peptides in LC or LC-MS applications. Each batch is QC tested.
  • Withstands 15kpsi operating pressure and handles pressure fluctuations during injection, providing superior longevity
  • 300 μm ID columns utilize an ultra-low dispersion column design
  • CSH particles provide excellent peak shape for peptides using FA- or TFA-containing mobile phases
  • Exhibits little dependence on strong ion pairing agents
  • Suggested uses include bottom-up proteomics, nano-flow, and trap and elute
Catalog No. 50-203-8950