Learn More
Promega NanoLuc-ULK1 Fusion Vector, 20microg
Used as a standard cloning vector, a template for in vitro transcription or for production of circular ssDNA.
Supplier:  Promega NV2211
Catalog No. PRNV2211
Add to cart
Description
Description
  • Standard Cloning Vectors That Can Produce ssDNA and In vitro Transcribe Sense and Antisense RNA
  • Blue/white screening for easy identification of recombinant clones
  • In vitro transcription using the SP6 and T7 RNA polymerase promoters flanking the multiple cloning region
  • The difference between + and − vectors is orientation of the f1 origin
Specifications
Specifications
20 μg
-30°C to -10°C