Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NANP Mouse anti-Human, Clone: OTI2C2, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA504557

Catalog No. 50-167-5215


Add to cart

Description

Description

NANP is a protein coding gene.
Specifications

Specifications

NANP
Monoclonal
1.37 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q8TBE9
C20orf147, HDHD4, dJ694B14.3
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
140838
Immunocytochemistry, Immunofluorescence, Western Blot
OTI2C2
Unconjugated
NANP
Liquid
NANP
Full length human recombit protein of human NANP produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit