Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAP1L1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550242

Catalog No. PA550242


Add to cart

Description

Description

The antibody detects endogenous levels of total NAP1L1 protein.

NAP1L1 is a member of the nucleosome assembly protein (NAP) family. This protein participates in DNA replication and may play a role in modulating chromatin formation and contribute to the regulation of cell proliferation.
Specifications

Specifications

NAP1L1
Polyclonal
Unconjugated
NAP1L1
Liquid
NAP1L1
Fusion protein corresponding to residues near the N terminal of human nucleosome assembly protein 1-like 1
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Immunohistochemistry (Paraffin), Western Blot
1.9 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
P55209, P28656
NRP, NAP1, NAP1L
Rabbit
IgG
100 μL
-20°C
Primary
4673, 53605
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit