Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAP1L1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549167

Catalog No. PA549167

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine, mouse and rat based on sequence homology.

NAP1L1 is a member of the nucleosome assembly protein (NAP) family. This protein participates in DNA replication and may play a role in modulating chromatin formation and contribute to the regulation of cell proliferation.
Specifications

Specifications

NAP1L1
Polyclonal
PBS with 0.09% sodium azide
P55209
Nucleosome assembly protein 1-like 1, NAP-1-related protein, hNRP, NAP1L1, NRP
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4673
Flow Cytometry, Western Blot
Unconjugated
NAP1L1
Liquid
NAP1L1
KLH conjugated synthetic peptide between 352-381 amino acids from the C-terminal region of human NAP1L1.
Ammonium sulfate precipitation
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit