Learn More
NAP1L1 Rabbit anti-Human, Unconjugated, Polyclonal, Active Motif™
Rabbit Polyclonal Antibody
Manufacturer:  Active MotifSupplier Diversity Partner 39841
Encompass
 View more versions of this product
Catalog No. 50-199-2897
Add to cart
Description
Description
NAP1L1 is a member of the nucleosome assembly protein (NAP) family. This protein participates in DNA replication and may play a role in modulating chromatin formation and contribute to the regulation of cell proliferation.
Specifications
Specifications
NAP1L1
Western Blot
Unconjugated
Nap1l1
Liquid
Rabbit
IgG
100 μL
-20°C, Avoid Freeze/Thaw Cycles
Primary
4673
Nap1l1
Lot-Specific
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
P55209
AA407126; AI256722; Brain protein DN38; D10Ertd68e; hNRP; HSP22-like protein interacting protein; NAP1; NAP-1; NAP1L; Nap1l1; NAP-1-related protein; NRP; nucleosome assembly protein 1 like 1; Nucleosome assembly protein 1-like 1; nucleosome assembly protein-1
This NAP1L1 antibody was raised against a peptide within the N-terminal region of NAP1L1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human