Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAP1L2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548987

Catalog No. PA548987

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse based on sequence homology.

Acidic protein which may be involved in interactions with other proteins or DNA.
Specifications

Specifications

NAP1L2
Polyclonal
PBS with 0.09% sodium azide
Q9ULW6
Nucleosome assembly protein 1-like 2, Brain-specific protein, X-linked, NAP1L2, BPX
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4674
Western Blot
Unconjugated
NAP1L2
Liquid
NAP1L2
KLH conjugated synthetic peptide between 113-139 amino acids from the N-terminal region of human NAP1L2.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit