Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAP1L3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548864

Catalog No. PA548864

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene is intronless and encodes a member of the nucleosome assembly protein (NAP) family. This gene is linked closely to a region of genes responsible for several X-linked mental retardation syndromes. [provided by RefSeq, Dec 2010]
Specifications

Specifications

NAP1L3
Polyclonal
PBS with 0.09% sodium azide
Q99457
Nucleosome assembly protein 1-like 3, NAP1L3, BNAP
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4675
Western Blot
Unconjugated
NAP1L3
Liquid
NAP1L3
KLH conjugated synthetic peptide between 432-459 amino acids from the C-terminal region of human NAP1L3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit