Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAP1L4 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$204.00 - $482.50
Specifications
Antigen | NAP1L4 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
NAP1L4 Polyclonal specifically detects NAP1L4 in Mouse samples. It is validated for Western Blot.Specifications
NAP1L4 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
4676 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Mouse | |
hNAP2, MGC4565, NAP-2, NAP2L, NAP2NAP1L4b, nucleosome assembly protein 1-like 4, nucleosome assembly protein 1-like 4b, Nucleosome assembly protein 2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-375 of human NAP1L4 (NP_005960.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title