Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAPA Mouse anti-Bovine, Clone: 15D4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP266628

Catalog No. NBP266628


Add to cart

Description

Description

NAPA Monoclonal antibody specifically detects NAPA in Bovine samples. It is validated for Western Blot, Immunoprecipitation, Enzyme Immunoassay.
Specifications

Specifications

NAPA
15D4
Western Blot, Immunoprecipitation 1:10 - 1:500, Enzyme Immunoassay
Affinity Purified
NAPA
Balb/C mice were immunized with recombinant human -SNAP. The resultant hybridomas were selected for reactivity with the immunogen by ELISA and with the antigen by western blot. The antibody was purified to >95% homogeneity by standard chromatographic techniques
Affinity purified
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
Bovine, Human, Rat
ELISA, Immunoprecipitation, Western Blot
Unconjugated
0.02M sodium phosphate, pH 7.5, 0.15M sodium chloride, and 50% glycerol with No Preservative
alpha-SNAP, alpha-soluble NSF attachment protein, N-ethylmaleimide-sensitive factor attachment protein alpha, N-ethylmaleimide-sensitive factor attachment protein, alpha, S, SNAPA, SNAP-alpha
Mouse
IgG1
100μg
Primary
8775
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit