Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAPA Mouse anti-Bovine, Clone: 16D1, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP266630
Description
NAPA Monoclonal antibody specifically detects NAPA in Bovine samples. It is validated for Western Blot.Specifications
NAPA | |
16D1 | |
Western Blot | |
Affinity Purified | |
NAPA | |
Recombinant human alpha SNAP | |
Affinity purified | |
Store at -20°C. Avoid freeze-thaw cycles. | |
Monoclonal | |
Bovine, Chicken, Human, Rat |
Western Blot | |
Unconjugated | |
20MM sodium phosphate, 150MM sodium chloride, 50% glycerol, pH 7.5 with No Preservative | |
alpha-SNAP, alpha-soluble NSF attachment protein, N-ethylmaleimide-sensitive factor attachment protein alpha, N-ethylmaleimide-sensitive factor attachment protein, alpha, S, SNAPA, SNAP-alpha | |
Mouse | |
IgG2b | |
100μg | |
Primary | |
8775 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title