Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

NAPE PLD Mouse anti-Human, Clone: OTI4B12, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525760

Catalog No. PIMA525760



Description

Description

N-Acylphosphatidylethanolamine (NAPE)-hydrolyzing phospholipase D (NAPE-PLD) is a membrane-bound enzyme which releases the endocannabinoid adamide and other bioactive N-acylethanolamines from their corresponding NAPEs in animal tissues. NAPE-PLD has been shown to be expressed by specific populations of neurons in the brain and targeted to axonal processes. Recent studies have suggested that NAEs generated by NAPE-PLD in axons may act as anterograde synaptic signaling molecules that regulate the activity of postsynaptic neurons. NAPE-PLD has also been shown to catalyze the hydrolysis of NAPE to generate OEA (oleoylethanolamide), a natural lipid amide that inhibits food intake in free-feeding rodents by prolonging latency to feed and postmeal interval.
Specifications

Specifications

NAPE PLD
Monoclonal
0.74 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q6IQ20
N-acyl phosphatidylethanolamine phospholipase D, DKFZp781D1098, FMP30, NAPE-PLD
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
222236
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI4B12
Unconjugated
NAPEPLD
Liquid
NAPEPLD
Full length human recombit NAPEPLD protein produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit