Alfa Aesar™ Naphthalene-1-sulfonyl chloride, 98%

Manufacturer: Alfa Aesar™ B2425006

Quantity 5g

 View more versions of this product

Catalog No. AAB2425006

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 85-46-1
    Molecular Formula C10H7ClO2S
    Formula Weight 226.68
    MDL Number MFCD00003984
    UN Number UN3261
    Beilstein 2099333
    Chemical Name or Material Naphthalene-1-sulfonyl chloride

    Specifications

    Formula Weight 226.68
    Quantity 5g
    TSCA TSCA
    Molecular Formula C10H7ClO2S
    MDL Number MFCD00003984
    CAS 85-46-1
    UN Number UN3261
    Beilstein 2099333
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Boiling Point 194° to 195°C (13mmHg)
    Melting Point 66° to 70°C
    Chemical Name or Material Naphthalene-1-sulfonyl chloride
    Assay 98%

    Description

    Applications
    1-Naphthalenesulfonyl chloride has been used in the preparation of 5 ft.-O-naphthaleneiulfonyldeoxyuridine and 1-sulfonylindazole derivatives. The derivatives of this compound is used for quantitative metabolome analysis New set of isotope reagents, 12C4-, 12C213C2-, and 13C4-5-diethylamino-naphthalene-1-sulfonyl chloride (DensCl), in combination with liquid chromatography Fourier-transform ion cyclotron resonance mass spectrometry (LC-FTICR-MS) is used for improved analysis of the amine- and phenol-containing submetabolome.

    Solubility
    Reacts slowly with water. Soluble in chloroform.

    Notes
    Hygroscopic. Store at dry place. Incompatible with oxidizing agents, bases and acids.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit